2-Phenylimidazo[1,2-a]pyridine-3-carboxylic acid, 98%
2-Phenylimidazo[1,2-a]pyridine-3-carboxylic acid, 98%
2-Phenylimidazo[1,2-a]pyridine-3-carboxylic acid, 98%
Thermo Scientific Chemicals

2-Phenylimidazo[1,2-a]pyridine-3-carboxylic acid, 98%

CAS: 123533-41-5 | C14H10N2O2 | 238.25 g/mol
製品番号(カタログ番号) H50236.MD
または、製品番号H50236-MD
価格(JPY)
-
数量:
250 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS123533-41-5
Assay (HPLC)98%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only