Isobutyramidoxime, 96%
Isobutyramidoxime, 96%
Isobutyramidoxime, 96%
Thermo Scientific Chemicals

Isobutyramidoxime, 96%

CAS: 35613-84-4 | C4H10N2O | 102.14 g/mol
製品番号(カタログ番号) H50892.03
または、製品番号H50892-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS35613-84-4
IUPAC Name(E)-N'-hydroxy-2-methylpropanimidamide
Molecular FormulaC4H10N2O
InChI KeyJHRDEHLFNLLCQS-UHFFFAOYSA-N
SMILESCC(C)C(N)=N/O
さらに表示
Melting Point (DSC)58.5-67.5?C
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥95.0 to ≤105.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only