Thermo Scientific Chemicals

Isobutyramidoxime, 96%, Thermo Scientific Chemicals

Catalog number: H50892.14
25 g, Each
Thermo Scientific Chemicals

Isobutyramidoxime, 96%, Thermo Scientific Chemicals

Catalog number: H50892.14
25 g, Each
Quantity
Catalog number: H50892.14
also known as H50892-14
Price (USD)
Price: 822.00
Online price: 711.65
Your price:
Quantity
-

Chemical Identifiers

CAS
35613-84-4
IUPAC Name
(E)-N'-hydroxy-2-methylpropanimidamide
Molecular Formula
C4H10N2O
InChI Key
JHRDEHLFNLLCQS-UHFFFAOYSA-N
SMILES
CC(C)C(N)=N/O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥95.0 to ≤105.0%
Melting Point (DSC)
58.5-67.5?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text