4-(4-Piperidinyloxy)benzonitrile, 98%
4-(4-Piperidinyloxy)benzonitrile, 98%
4-(4-Piperidinyloxy)benzonitrile, 98%
Thermo Scientific Chemicals

4-(4-Piperidinyloxy)benzonitrile, 98%

CAS: 224178-67-0 | C12H14N2O | 202.257 g/mol
製品番号(カタログ番号) H50958.MD
または、製品番号H50958-MD
価格(JPY)
-
数量:
250 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS224178-67-0
IUPAC Name4-(piperidin-4-yloxy)benzonitrile
Molecular FormulaC12H14N2O
InChI KeyDRIREFRFHFBPIN-UHFFFAOYSA-N
SMILESN#CC1=CC=C(OC2CCNCC2)C=C1
さらに表示
Assay (HPLC)≥97.5%
FormSolid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only