1-Aza-15-crown-5, 97%
1-Aza-15-crown-5, 97%
1-Aza-15-crown-5, 97%
Thermo Scientific Chemicals

1-Aza-15-crown-5, 97%

CAS: 66943-05-3 | C10H21NO4 | 219.281 g/mol
数量:
250 mg
1 g
5 g
製品番号(カタログ番号) H51064.MD
または、製品番号H51064-MD
価格(JPY)
-
数量:
250 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS66943-05-3
IUPAC Name1,4,7,10-tetraoxa-13-azacyclopentadecan-13-ium
Molecular FormulaC10H22NO4
InChI KeyBJUOQSZSDIHZNP-UHFFFAOYSA-O
SMILESC1COCCOCCOCCOCC[NH2+]1
さらに表示
CommentConverts to solid at low temperature
Appearance (Color)Pale yellow
Assay by NMR>96.0%
FormOil
1-Aza-15-crown-5 is used as building block for the synthesis of bis-crown ethers, lariat ethers and other monofuctionalized macrocycles. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Aza-15-crown-5 is used as building block for the synthesis of bis-crown ethers, lariat ethers and other monofuctionalized macrocycles. It is also used as pharmaceutical intermediate.

Solubility
Soluble in water and methanol.

Notes
Air sensitive. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Pu Qian; Toshiro Yokoyama; Osamu Itabashi; Tomio Goto; Tokuji Miyashita. Ag+ ion sensing by the polymer Langmuir-Blodgett films containing 1-aza-15-crown 5-ether group. Thin Solid Films. 1997, 305, (1-2),316-320
  2. Kh. A.Hassan. Spectroscopic studies of complexes of iodine with the bases 1-aza-15-crown-5 and 3,6,9,14-tetrathiabicyclo[9.2.1]tetradeca-11,13-diene. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2004, 60, (13),3059-3063