4-(Tri-n-butylstannyl)thiazole, 95%
4-(Tri-n-butylstannyl)thiazole, 95%
4-(Tri-n-butylstannyl)thiazole, 95%
Thermo Scientific Chemicals

4-(Tri-n-butylstannyl)thiazole, 95%

CAS: 173979-01-6 | C15H29NSSn | 374.17 g/mol
製品番号(カタログ番号) H51457.MD
または、製品番号H51457-MD
価格(JPY)
-
数量:
250 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS173979-01-6
FormLiquid
Assay from Supplier's CofA≥94.0%
4-(Tri-n-butylstannyl)thiazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Tri-n-butylstannyl)thiazole is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Rudiger Wittenberg; Jiri Srogl; Masahiro Egi; Lanny S.Liebeskind. Ketone synthesis under neutral conditions. Cu(I) diphenylphosphinate-mediated, palladium-catalyzed coupling of thiol esters and organostannanes. Org. Lett. 2003, 5, (17),3033-3035
  2. Hao Li; Hao Yang; Lanny S.Liebeskind. Synthesis of high enantiopurity N-Protected α-amino ketones by thiol ester-Organostannane cross-coupling using pH-neutral conditions. Org. Lett. 2008, 10, (19),4375-4378