2-Chloro-1-tritylimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloro-1-tritylimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only
Kenneth L.Kirk. Facile synthesis of 2-substituted imidazoles. J. Org. Chem. 1978, 43, (22),4381-4383