2-Chloro-1-methylimidazole, 97+%
2-Chloro-1-methylimidazole, 97+%
2-Chloro-1-methylimidazole, 97+%
Thermo Scientific Chemicals

2-Chloro-1-methylimidazole, 97+%

CAS: 253453-91-7 | C4H5ClN2 | 116.55 g/mol
製品番号(カタログ番号) H51867.MD
または、製品番号H51867-MD
価格(JPY)
-
数量:
250 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS253453-91-7
Assay (unspecified)≥97.0% (GC)
Proton NMRConforms to structure
Appearance (Color)Colorless to pale brown
FormLiquid or viscous liquid
Assay≥97.0% (GC)
2-Chloro-1-methylimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-1-methylimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
E.V.Aleksandrova; A.N.Kravchenko; P.M.Kochergin. Methods for the synthesis of haloimidazoles (review). Chemistry of Heterocyclic Compounds. 2011, 46, (11),1295-1317