4-(Diphenylamino)benzeneboronic acid pinacol ester, 95%
4-(Diphenylamino)benzeneboronic acid pinacol ester, 95%
4-(Diphenylamino)benzeneboronic acid pinacol ester, 95%
Thermo Scientific Chemicals

4-(Diphenylamino)benzeneboronic acid pinacol ester, 95%

CAS: 267221-88-5 | C24H26BNO2 | 371.29 g/mol
製品番号(カタログ番号) H51946.03
または、製品番号H51946-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
仕様
バイルシュタイン10571706
CAS267221-88-5
化学物質名または材質4-(Diphenylamino)benzeneboronic acid pinacol ester
式量371.29
IUPAC名N,N-diphenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only