N-Boc-D-pipecolinic acid, 98%
N-Boc-D-pipecolinic acid, 98%
N-Boc-D-pipecolinic acid, 98%
N-Boc-D-pipecolinic acid, 98%
Thermo Scientific Chemicals

N-Boc-D-pipecolinic acid, 98%

CAS: 28697-17-8 | C11H19NO4 | 229.276 g/mol
製品番号(カタログ番号) H51961.14
または、製品番号H51961-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS28697-17-8
IUPAC Name1-[(tert-butoxy)carbonyl]piperidine-2-carboxylic acid
Molecular FormulaC11H19NO4
InChI KeyJQAOHGMPAAWWQO-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CCCCC1C(O)=O
さらに表示
Optical Rotation+55.0° to +65.0° (C₁, AcOH)
FormPowder
Appearance (Color)White
Assay (HPLC)≥97.5%
It is also applied as catalytic agent and petrochemical additive. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is also applied as catalytic agent and petrochemical additive. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

Solubility
Soluble in 2ml dimethyl formamide 0.3gram.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
Andrew G. Myers, et al. Practical method for the synthesis of D- or L-.alpha.-amino acids by the alkylation of (+)- or (-)-pseudoephedrine glycinamide.J. Am. Chem. Soc.,1995,117(32), 8488-8489.