(S)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid, 98%
(S)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid, 98%
(S)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid, 98%
Thermo Scientific Chemicals

(S)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid, 98%

CAS: 122235-70-5 | C23H26N2O6 | 426.469 g/mol
製品番号(カタログ番号) H51964.03
または、製品番号H51964-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS122235-70-5
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Molecular FormulaC23H26N2O6
InChI KeyMVWPBNQGEGBGRF-IBGZPJMESA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
さらに表示
Appearance (Color)White
Infrared spectrumConforms
Purity≥97.5% (HPLC)
FormPowder
It has been used in the synthesis of target chelator, the tris-catechol compound 2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been used in the synthesis of target chelator, the tris-catechol compound 2.

Solubility
Soluble in 2ml dimethyl formamide 0.3gram.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
Etienne Baco, et al. Diphenyl-benzo[1,3]dioxole-4-carboxylic acid pentafluorophenyl ester: a convenient catechol precursor in the synthesis of siderophore vectors suitable for antibiotic Trojan horse strategies.Org. Biomol. Chem.,2014,12(5), 749-757.