4-Amino-N-Boc-L-phenylalanine, 95%
4-Amino-N-Boc-L-phenylalanine, 95%
4-Amino-N-Boc-L-phenylalanine, 95%
Thermo Scientific Chemicals

4-Amino-N-Boc-L-phenylalanine, 95%

CAS: 55533-24-9 | C14H20N2O4 | 280.32 g/mol
製品番号(カタログ番号) H51980.03
または、製品番号H51980-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS55533-24-9
IUPAC Name(2S)-3-(4-aminophenyl)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Molecular FormulaC14H20N2O4
InChI KeyNDMVQEZKACRLDP-NSHDSACASA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CC1=CC=C(N)C=C1)C(O)=O
さらに表示
Optical Rotation|b = +25? ?2? (C=1 in MeOH)
Solution TestClear solution (0.3 g in 2mL DMF)
Appearance (Color)White to pale cream
Assay (HPLC)≥94.0%
Assay from Supplier's CofAMass Spec conforms
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only