6-Bromo-1-tetralone, 96%, Thermo Scientific Chemicals
6-Bromo-1-tetralone, 96%, Thermo Scientific Chemicals
6-Bromo-1-tetralone, 96%, Thermo Scientific Chemicals
6-Bromo-1-tetralone, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Bromo-1-tetralone, 96%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
H52381.03
または、製品番号H52381-03
1 g
製品番号(カタログ番号) H52381.03
または、製品番号H52381-03
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
仕様
CAS66361-67-9
化学物質名または材質6-Bromo-1-tetralone
式量225.09
IUPAC名6-bromo-3,4-dihydro-2H-naphthalen-1-one
InChIキーOSDHOOBPMBLALZ-UHFFFAOYSA-N
さらに表示
6-bromo-1-tetralone used in Wittig-Horner reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-bromo-1-tetralone used in Wittig-Horner reaction.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, heat.
RUO – Research Use Only
Munefumi Kanao,; Yoshifumi Watanabe,; Youichi Kimura,; Junji Saegusa,; Kenjiro Yamamoto,; Hideyuki Kanno,; Naoaki Kanaya,; Hideo Kubo,; Shinichiro Ashida,; Fumiyoshi Ishikawa.Thromboxane A2 synthetase inhibitors. 2. Syntheses and activities of tetrahydronaphthalene and indane derivatives. J. Med. Chem., 1999, 32 (6), 1326-1334.