N-Boc-2-methyl-L-serine, 97%
N-Boc-2-methyl-L-serine, 97%
N-Boc-2-methyl-L-serine, 97%
Thermo Scientific Chemicals

N-Boc-2-methyl-L-serine, 97%

CAS: 84311-19-3 | C9H17NO5 | 219.24 g/mol
製品番号(カタログ番号) H52570.03
または、製品番号H52570-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
仕様
CAS84311-19-3
化学物質名または材質N-Boc-2-methyl-L-serine
式量219.24
IUPAC名(2S)-3-hydroxy-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
InChIキーFWRXDSRYWWYTPD-VIFPVBQESA-N
さらに表示
N-Boc-2-methyl-L-serine is used as a cocatalyst in the Diels-Alder reactions between cyclopentadiene and methyl acrylate in the presence of Lewis acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-2-methyl-L-serine is used as a cocatalyst in the Diels-Alder reactions between cyclopentadiene and methyl acrylate in the presence of Lewis acids.

Solubility
Soluble in dimethylformamide.

Notes
Incompatible with reducing agents, water and strong oxidizing agents.
RUO – Research Use Only
Evindar, G.; Satz, A. L.; Bernier, S. G.; Kavarana, M. J.; Doyle, E.; Lorusso, J.; Taghizadeh, N.; Halley, K.; Hutchings, A.; Kelley, M. S.; Wright, A. D. Synthesis and evaluation of arylalkoxy- and biarylalkoxy- phenylamide and phenylimidazoles as potent and selective sphingosine-1-phosphate receptor subtype-1 agonists. Bioorg. Med. Chem. Lett. 2009, 19 (8), 2315-2319.