1-Boc-5-methoxyindole-2-boronic acid, 95%
1-Boc-5-methoxyindole-2-boronic acid, 95%
1-Boc-5-methoxyindole-2-boronic acid, 95%
Thermo Scientific Chemicals

1-Boc-5-methoxyindole-2-boronic acid, 95%

CAS: 290331-71-4 | C14H18BNO5 | 291.11 g/mol
製品番号(カタログ番号) H52654.03
または、製品番号H52654-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS290331-71-4
Appearance (Color)White to pale cream or pale yellow
FormPowder or crystalline powder or crystals and/or chunks
Assay by NMR≥94.0% (proton-NMR)
Assay from Suppliers CofA>94.0% (HPLC)
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only