(3-Methylsulfonylaminomethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals
(3-Methylsulfonylaminomethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals
(3-Methylsulfonylaminomethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(3-Methylsulfonylaminomethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
H52705.03
または、製品番号H52705-03
1 g
製品番号(カタログ番号) H52705.03
または、製品番号H52705-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS850568-39-7
IUPAC Name[3-(methanesulfonamidomethyl)phenyl]boronic acid
Molecular FormulaC8H12BNO4S
InChI KeyFJSWJJOLFPIDER-UHFFFAOYSA-N
SMILESCS(=O)(=O)NCC1=CC=CC(=C1)B(O)O
さらに表示
Assay (HPLC)>97.5%
(3-Methylsulfonylaminomethyl)benzeneboronic acid, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Methylsulfonylaminomethyl)benzeneboronic acid, is used as a pharmaceutical intermediate.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions
RUO – Research Use Only