4-(Boc-aminomethyl)piperidine is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Boc-aminomethyl)piperidine is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in DMSO.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
- V Veljkovic.; N Veljkovic.; JA Este. Application of the EIIP/ISM bioinformatics concept in development of new drugs. Current Medicinal Chemistry. 200714 (4), 441-453.
- N Langlois. Diastereoselective synthesis of enantiopure differentially protected cis-4, 5-diaminopiperidin-2-one through intramolecular transamidation. Tetrahedron letters. 200243 (52), 9531-9533.