N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine, 97%
N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine, 97%
N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine, 97%
Thermo Scientific Chemicals

N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine, 97%

CAS: 85326-10-9 | C20H33N5O5Si | 451.599 g/mol
製品番号(カタログ番号) H52726.03
または、製品番号H52726-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
仕様
CAS85326-10-9
化学物質名または材質N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine
式量451.6
IUPAC名N-[9-[(2R,4S,5R)-5-[[tert-butyl(dimethyl)silyl]oxymethyl]-4-hydroxyoxolan-2-yl]-6-oxo-3H-purin-2-yl]-2-methylpropanamide
InChIキーWZQWRHWLQPEBFF-BFHYXJOUSA-N
さらに表示
N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Isobutyryl-5′-O-tert-butyldimethylsilyl-2′-deoxyguanosine is used as pharmaceutical intermediate.

Notes
Moisture sensitive. Store away from oxidizing agents.
RUO – Research Use Only