trans-N-Boc-4-cyano-L-proline methyl ester, 97%, Thermo Scientific Chemicals
trans-N-Boc-4-cyano-L-proline methyl ester, 97%, Thermo Scientific Chemicals
trans-N-Boc-4-cyano-L-proline methyl ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-N-Boc-4-cyano-L-proline methyl ester, 97%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
H52733.03
または、製品番号H52733-03
1 g
製品番号(カタログ番号) H52733.03
または、製品番号H52733-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS194163-91-2
IUPAC Name1-tert-butyl 2-methyl 4-cyanopyrrolidine-1,2-dicarboxylate
Molecular FormulaC12H18N2O4
InChI KeyGHLKJIQVORAVHE-UHFFFAOYNA-N
SMILESCOC(=O)C1CC(CN1C(=O)OC(C)(C)C)C#N
さらに表示
Optical purity98.0% min
Assay (unspecified)96.0% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only