(R)-3-(Boc-amino)piperidine, 97%
(R)-3-(Boc-amino)piperidine, 97%
(R)-3-(Boc-amino)piperidine, 97%
Thermo Scientific Chemicals

(R)-3-(Boc-amino)piperidine, 97%

CAS: 309956-78-3 | C10H20N2O2 | 200.282 g/mol
製品番号(カタログ番号) H52749.06
または、製品番号H52749-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS309956-78-3
IUPAC Nametert-butyl N-[(3R)-piperidin-3-yl]carbamate
Molecular FormulaC10H20N2O2
InChI KeyWUOQXNWMYLFAHT-MRVPVSSYSA-N
SMILESCC(C)(C)OC(=O)N[C@@H]1CCCNC1
さらに表示
Appearance (Color)White to pale yellow
FormSolid
Assay (unspecified)>96.0%
Loss on Drying<0.50%
Optical purity>98.0%
さらに表示
(R)-3-(Boc-amino)piperidine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-3-(Boc-amino)piperidine is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol and ethanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Jeremy N.Burrows; John G.Cumming; Shaun M.Fillery; Gordon A.Hamlin; Julian A.Hudson; Ruth J.Jackson; Sharon McLaughlin; John S.Shaw. Modulators of the human CCR5 receptor. Part 1: Discovery and initial SAR of 1-(3,3-diphenylpropyl)-piperidinyl amides and ureas. Bioorganic & Medicinal Chemistry Letters. 2005, 15, (1),25-28