(S)-3-Amino-1-benzylpiperidine, 97%
(S)-3-Amino-1-benzylpiperidine, 97%
(S)-3-Amino-1-benzylpiperidine, 97%
Thermo Scientific Chemicals

(S)-3-Amino-1-benzylpiperidine, 97%

CAS: 168466-85-1 | C12H18N2 | 190.29 g/mol
製品番号(カタログ番号) H52772.03
または、製品番号H52772-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
仕様
CAS168466-85-1
化学物質名または材質(S)-3-Amino-1-benzylpiperidine
式量190.29
健康被害1H315-H319-H335
健康被害2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
さらに表示
(S)-3-Amino-1-benzylpiperidine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-3-Amino-1-benzylpiperidine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Freely soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only
J Moragues.; J Prieto.; RGW Spickett. Rearrangements during the synthesis of substituted 1-benzylpyrrolidines and 3-substituted 1-benzylpiperidines. J. Chem. Soc., Perkin Trans. 1. 19761 (9), 938-940.