(1S,2S)-2-Aminocyclohexanecarboxylic acid, 97%
(1S,2S)-2-Aminocyclohexanecarboxylic acid, 97%
(1S,2S)-2-Aminocyclohexanecarboxylic acid, 97%
Thermo Scientific Chemicals

(1S,2S)-2-Aminocyclohexanecarboxylic acid, 97%

CAS: 24716-93-6 | C7H13NO2 | 143.186 g/mol
数量:
250 mg
1 g
5 g
製品番号(カタログ番号) H52779.06
または、製品番号H52779-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
仕様
CAS24716-93-6
化学物質名または材質(1S,2S)-2-Aminocyclohexanecarboxylic acid
式量143.19
健康被害1H315-H319-H335
健康被害2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
さらに表示
(1S,2S)-2-Aminocyclohexanecarboxylic acid is used in the preparation of enantiomerically pure beta-amino acids. It is also involved in the synthesis of mono and dihydroxylated derivatives of 2-aminocyclohexanecarboxylic acid by Diels-Alder adduct of ethyl(E)-3-nitroacrylate and furan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1S,2S)-2-Aminocyclohexanecarboxylic acid is used in the preparation of enantiomerically pure beta-amino acids. It is also involved in the synthesis of mono and dihydroxylated derivatives of 2-aminocyclohexanecarboxylic acid by Diels-Alder adduct of ethyl(E)-3-nitroacrylate and furan.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Vilaivan, T. Pyrrolidinyl PNA with alpha/beta-Dipeptide Backbone: From Development to Applications. Acc. Chem. Res. 2015, 48 (6), 1645-1656.
  2. Ma, C. D.; Wang, C.; Acevedo-Vélez, C.; Gellman, S. H.; Abbott, N. L. Modulation of hydrophobic interactions by proximally immobilized ions. Nature 2015, 517 (7534), 347-350.