3-Aminobenzeneboronic acid, 98%
3-Aminobenzeneboronic acid, 98%
3-Aminobenzeneboronic acid, 98%
Thermo Scientific Chemicals

3-Aminobenzeneboronic acid, 98%

CAS: 30418-59-8 | C6H8BNO2 | 136.945 g/mol
製品番号(カタログ番号) H52909.03
または、製品番号H52909-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS30418-59-8
IUPAC Name(3-aminophenyl)boronic acid
Molecular FormulaC6H8BNO2
InChI KeyJMZFEHDNIAQMNB-UHFFFAOYSA-N
SMILESNC1=CC=CC(=C1)B(O)O
さらに表示
Assay (HPLC)>97.5%
3-Aminobenzeneboronic acid is used as an organic chemical synthesis intermediate. It is a boronic acid with potential use for biochemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminobenzeneboronic acid is used as an organic chemical synthesis intermediate. It is a boronic acid with potential use for biochemical research.

Solubility
Soluble in water (partly miscible).

Notes
Stable under recommended storage conditions. Store away from oxidizing agents and heat.
RUO – Research Use Only

General References:

  1. David A.Widdowson; Rene Wilhelm. Palladium catalysed Suzuki reactions of fluoroarenes. Chem. Commun. 2003, (5),578-579
  2. James H.Hageman; Glenn D.Kuehn. Boronic acid matrices for the affinity purification of glycoproteins and enzymes. Practical Protein Chromatography. 1992, 11, 45-71