3-Fluoropyridine-4-boronic acid, 98%
3-Fluoropyridine-4-boronic acid, 98%
3-Fluoropyridine-4-boronic acid, 98%
Thermo Scientific Chemicals

3-Fluoropyridine-4-boronic acid, 98%

CAS: 458532-97-3 | C5H5BFNO2 | 140.908 g/mol
製品番号(カタログ番号) H53109.03
または、製品番号H53109-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS458532-97-3
IUPAC Name(3-fluoropyridin-4-yl)boronic acid
Molecular FormulaC5H5BFNO2
InChI KeyQUHSESYLMZVXCN-UHFFFAOYSA-N
SMILESOB(O)C1=CC=NC=C1F
さらに表示
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only