1-Phenylvinylboronic acid pinacol ester, 95%, Thermo Scientific Chemicals
1-Phenylvinylboronic acid pinacol ester, 95%, Thermo Scientific Chemicals
1-Phenylvinylboronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Phenylvinylboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
H53216.03
または、製品番号H53216-03
1 g
製品番号(カタログ番号) H53216.03
または、製品番号H53216-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
仕様
CAS143825-84-7
化学物質名または材質1-Phenylvinylboronic acid pinacol ester
式量230.11
IUPAC名4,4,5,5-tetramethyl-2-(1-phenylethenyl)-1,3,2-dioxaborolane
InChIキーRMGBWPMWUZSIMH-UHFFFAOYSA-N
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only