6-Chloro-1-hexene, 97%
6-Chloro-1-hexene, 97%
6-Chloro-1-hexene, 97%
Thermo Scientific Chemicals

6-Chloro-1-hexene, 97%

CAS: 928-89-2 | C6H11Cl | 118.604 g/mol
製品番号(カタログ番号) H53396.14
または、製品番号H53396-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
仕様
CAS928-89-2
化学物質名または材質6-Chloro-1-hexene
DOT情報Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
密度0.896
EINECS番号213-186-5
さらに表示
6-Chloro-1-hexene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloro-1-hexene is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Miguel Yus; Rosa Ortiz; Fernando F.Huerta. DTBB-catalysed lithiation of 6-chloro-1-hexene and related systems: synthetically useful temperature-dependent behaviour. Tetrahedron Letters. 2002, 43, (16),2957-2960
  2. Leonardus W.Jenneskens; Cornelis A.M.Hoefs; Ulfert E.Wiersum. Preparative flash vacuum thermolysis. Selective elimination of 6-chloro-1-hexene from esters of 6-chloro-1-hexanol with Schoenberg rearrangement of the S-methyl xanthate. J. Org. Chem. 1989, 54, (24),5811-5814