5-Decyn-1-ol, 97%
5-Decyn-1-ol, 97%
5-Decyn-1-ol, 97%
Thermo Scientific Chemicals

5-Decyn-1-ol, 97%

CAS: 68274-97-5 | C10H18O | 154.25 g/mol
製品番号(カタログ番号) H53400.14
または、製品番号H53400-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
仕様
CAS68274-97-5
化学物質名または材質5-Decyn-1-ol
密度0.861
EINECS番号269-529-4
式量154.25
さらに表示
5-Decyn-1-ol is used in the synthesis of multi-substituted cyclopropanes. It involved in manufacture of fine chemicals, having applications in electroplating and perfumery industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Decyn-1-ol is used in the synthesis of multi-substituted cyclopropanes. It involved in manufacture of fine chemicals, having applications in electroplating and perfumery industry.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Pei Nian Liu Dr.,Fu Hai Su Dr.,; Ting Bin Wen Dr.,; Herman H.-Y. Sung Dr.,; Ian D. Williams Prof. Dr., Guochen Jia Prof. Dr. Selective and Efficient Cycloisomerization of Alkynols Catalyzed by a New Ruthenium Complex with a Tetradentate Nitrogen-Phosphorus Mixed Ligand. TOC 2010, 16 (26) ,7889-7897.