4,6-Decadiyne, 97%
4,6-Decadiyne, 97%
4,6-Decadiyne, 97%
Thermo Scientific Chemicals

4,6-Decadiyne, 97%

CAS: 16387-71-6 | C10H14 | 134.222 g/mol
製品番号(カタログ番号) H53467.14
または、製品番号H53467-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS16387-71-6
IUPAC Namedeca-4,6-diyne
Molecular FormulaC10H14
InChI KeyLIWZSNTUMSGWTF-UHFFFAOYSA-N
SMILESCCCC#CC#CCCC
さらに表示
Appearance (Color)Colorless to yellow to brown
FormLiquid
Assay from Supplier's CofA≥96.0%
6-Decen-6-yne is prepared from 4,6-Decadiyne. PDA(poly 4,6-Decadiyne) is used in preparation of thin films. 4,6-decadiyne is used as a substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Decen-6-yne is prepared from 4,6-Decadiyne. PDA(poly 4,6-Decadiyne) is used in preparation of thin films. 4,6-decadiyne is used as a substrate.

Solubility
Immiscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Hirohisa Ohmiya,; Hideki Yorimitsu Dr.,; Koichiro Oshima Prof. Dr. Cobalt-Catalyzed syn Hydrophosphination of Alkynes . TOC 2005, 117(16), 2420-2422.