3-Methoxyquinoline-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals
3-Methoxyquinoline-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals
3-Methoxyquinoline-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methoxyquinoline-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
H54211.03
または、製品番号H54211-03
1 g
製品番号(カタログ番号) H54211.03
または、製品番号H54211-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
仕様
CAS881656-35-5
化学物質名または材質3-Methoxyquinoline-5-boronic acid pinacol ester
式量285.15
IUPAC名3-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
InChIキーMGEKTRDPBDREKY-UHFFFAOYSA-N
さらに表示
3-Methoxyquinoline-5-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxyquinoline-5-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Timo Leermann; Frederic R.Leroux; Francoise Colobert. Highly efficient one-pot access to functionalized arylboronic acids via noncryogenic bromine/magnesium exchanges. Org. Lett. 2011, 13, (17),4479-4481