1-(4-Methoxybenzyl)piperazine, 97%
1-(4-Methoxybenzyl)piperazine, 97%
1-(4-Methoxybenzyl)piperazine, 97%
Thermo Scientific Chemicals

1-(4-Methoxybenzyl)piperazine, 97%

CAS: 21867-69-6 | C12H20N2O | 208.30 g/mol
製品番号(カタログ番号) H55099.03
または、製品番号H55099-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
仕様
CAS21867-69-6
化学物質名または材質1-(4-Methoxybenzyl)piperazine
White to Yellow
引火点>110°C (230°F)
式量206.29
さらに表示
1-(4-Methoxybenzyl)piperazine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(4-Methoxybenzyl)piperazine is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. M.Teresa Bartolome.; Felix L. Buenadicha.; Carmen Avendano.; Mónica Söllhuber. Stereochemical course of acylation and aldol condensation in (4S)-4-methyl-2-benzyl-2,4-dihydro-1H-pyrazino[2,1-b]quinazoline-3,6-diones. Tetrahedron: Asymmetry. 1998, 9 (2), 249-258.
  2. Umide Demir Ozkay.; Ozgür Devrim Can.; Yusuf Ozkay.; Yusuf Ozturk. Effect of benzothiazole/piperazine derivatives on intracerebroventricular streptozotocin-induced cognitive deficits. Chemical Communications. 2012, 64 (4), 834-847.