Thermo Scientific Chemicals

S-Phenyl thioacetate, 98%, Thermo Scientific Chemicals

Catalog number: H55175.06
5 g, Each
Thermo Scientific Chemicals

S-Phenyl thioacetate, 98%, Thermo Scientific Chemicals

Catalog number: H55175.06
5 g, Each
Quantity
Catalog number: H55175.06
also known as H55175-06
Price (USD)
Price: 46.50
Online price: 39.65
Your price:
Quantity
-

Chemical Identifiers

CAS
934-87-2
IUPAC Name
1-(phenylsulfanyl)ethan-1-one
Molecular Formula
C8H8OS
InChI Key
WBISVCLTLBMTDS-UHFFFAOYSA-N
SMILES
CC(=O)SC1=CC=CC=C1
Appearance (Color)
Colorless to yellow
Assay from Suppliers CofA
≥97.5%
Form
Liquid or low melting point solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text