S-Phenyl thioacetate, 98%
S-Phenyl thioacetate, 98%
S-Phenyl thioacetate, 98%
S-Phenyl thioacetate, 98%
Thermo Scientific Chemicals

S-Phenyl thioacetate, 98%

CAS: 934-87-2 | C8H8OS | 152.211 g/mol
製品番号(カタログ番号) H55175.14
または、製品番号H55175-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS934-87-2
IUPAC Name1-(phenylsulfanyl)ethan-1-one
Molecular FormulaC8H8OS
InChI KeyWBISVCLTLBMTDS-UHFFFAOYSA-N
SMILESCC(=O)SC1=CC=CC=C1
さらに表示
Appearance (Color)Colorless to yellow
Assay from Suppliers CofA≥97.5%
FormLiquid or low melting point solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only