Agmatine sulfate, 97%
Agmatine sulfate, 97%
Agmatine sulfate, 97%
Thermo Scientific Chemicals

Agmatine sulfate, 97%

CAS: 2482-00-0 | C5H16N4O4S | 228.267 g/mol
数量:
250 mg
1 g
5 g
製品番号(カタログ番号) H55363.03
または、製品番号H55363-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2482-00-0
IUPAC NameN''-(4-aminobutyl)guanidine; sulfuric acid
Molecular FormulaC5H16N4O4S
InChI KeyPTAYFGHRDOMJGC-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.NCCCCN=C(N)N
さらに表示
Assay (HPLC)≥96.0% [US]
CommentMaterial sourced in either [US] or [UK]
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0% [UK]
FormPowder
Agmatine is an intermediate in the biosynthesis of the polyamines putrescine and spermine. Agmatine binds with high affinity to α2-adrenergic receptors and imidazoline binding sites and produces blockade of the NMDA-receptor associated cation channels. Agmatine is a competitive inhibitor of nitric oxide synthase activity owing to mimicry of the natural NOS substrate arginine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agmatine is an intermediate in the biosynthesis of the polyamines putrescine and spermine. Agmatine binds with high affinity to α2-adrenergic receptors and imidazoline binding sites and produces blockade of the NMDA-receptor associated cation channels. Agmatine is a competitive inhibitor of nitric oxide synthase activity owing to mimicry of the natural NOS substrate arginine.

Solubility
Soluble in water, nearly insoluble in alcohol

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Yoko Sakakibara.; Hiroshi Yanagisawa. Agmatine deiminase from cucumber seedlings is a mono-specific enzyme: purification and characteristics. Protein Expression and Purification. 2003, 30, (1), 88-93.
  2. Kazuho Abe.; Yuzuru Abe.; Hiroshi Saito. Agmatine suppresses nitric oxide production in microglia. Brain Research. 2000, 872, (1-2), 141-148.