4-(Boc-amino)phenol, 97%
4-(Boc-amino)phenol, 97%
4-(Boc-amino)phenol, 97%
4-(Boc-amino)phenol, 97%
Thermo Scientific Chemicals

4-(Boc-amino)phenol, 97%

CAS: 54840-15-2 | C11H15NO3 | 209.245 g/mol
製品番号(カタログ番号) H55645.14
または、製品番号H55645-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS54840-15-2
IUPAC Nametert-butyl N-(4-hydroxyphenyl)carbamate
Molecular FormulaC11H15NO3
InChI KeyYRQMBQUMJFVZLF-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NC1=CC=C(O)C=C1
さらに表示
Assay (HPLC)>96.0%
4-(Boc-amino)phenol, is used to protect amine in the solid phase synthesis of peptides, used to produce [4-(tert-butyl-dimethyl-silanyloxy)-phenyl]-carbamic acid tert-butyl ester at ambient temperature. This reaction will need reagent imidazole and solvent dimethylformamide, CH2Cl2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Boc-amino)phenol, is used to protect amine in the solid phase synthesis of peptides, used to produce [4-(tert-butyl-dimethyl-silanyloxy)-phenyl]-carbamic acid tert-butyl ester at ambient temperature. This reaction will need reagent imidazole and solvent dimethylformamide, CH2Cl2.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only