1-(4-Methoxyphenyl)piperazine, 97%
1-(4-Methoxyphenyl)piperazine, 97%
1-(4-Methoxyphenyl)piperazine, 97%
1-(4-Methoxyphenyl)piperazine, 97%
Thermo Scientific Chemicals

1-(4-Methoxyphenyl)piperazine, 97%

CAS: 38212-30-5 | C11H16N2O | 192.26 g/mol
製品番号(カタログ番号) H55791.03
または、製品番号H55791-03
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS38212-30-5
IUPAC Name1-(4-methoxyphenyl)piperazine
Molecular FormulaC11H16N2O
InChI KeyMRDGZSKYFPGAKP-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)N1CCNCC1
さらに表示
Assay (HPLC)>96.0%
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
soluble in water, methanol and toluene

Notes
Air Sensitive. Store under nitrogen.
RUO – Research Use Only

General References:

  1. Roland F. Staack.; Hans H. Maurer. Toxicological detection of the new designer drug 1-(4-methoxyphenyl)piperazine and its metabolites in urine and differentiation from an intake of structurally related medicaments using gas chromatography-mass spectrometry. J. Chromatogr. B. 2003, 798 (2),333-342.
  2. Douwe de Boer.; Ingrid J Bosman.; Elöd Hidvégi.; Carmo Manzoni.; András A Benkö.; Lourenço J.A.L dos Reys.; Robert A.A Maes. Piperazine-like compounds: a new group of designer drugs-of-abuse on the European market. Forensic Science International. 2001, 121 (1-2),47-56.