(R)-(+)-1-(2-Bromophenyl)ethanol, 98%
(R)-(+)-1-(2-Bromophenyl)ethanol, 98%
(R)-(+)-1-(2-Bromophenyl)ethanol, 98%
Thermo Scientific Chemicals

(R)-(+)-1-(2-Bromophenyl)ethanol, 98%

CAS: 76116-20-6 | C8H9BrO | 201.063 g/mol
製品番号(カタログ番号) H56003.06
または、製品番号H56003-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS76116-20-6
IUPAC Name(1R)-1-(2-bromophenyl)ethan-1-ol
Molecular FormulaC8H9BrO
InChI KeyDZLZSFZSPIUINR-ZCFIWIBFSA-N
SMILESC[C@@H](O)C1=CC=CC=C1Br
さらに表示
Assay (unspecified)>97.5%
It is the most important dyestuff intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is the most important dyestuff intermediate.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
RUO – Research Use Only
Morris Srebnik.; P. V. Ramachandran.; Herbert C. Brown. Chiral synthesis via organoboranes. 18. Selective reductions. 43. Diisopinocampheylchloroborane as an excellent chiral reducing reagent for the synthesis of halo alcohols of high enantiomeric purity. A highly enantioselective synthesis of both optical isomers of Tomoxetine, Fluoxetine, and Nisoxetine.J. Org. Chem. 1988, 53 (13),2916-2920 .