Thermo Scientific Chemicals

Azetidine, 98%, Thermo Scientific Chemicals

Catalog number: H56351.03
1 g, Each
Thermo Scientific Chemicals

Azetidine, 98%, Thermo Scientific Chemicals

Catalog number: H56351.03
1 g, Each
Quantity
Catalog number: H56351.03
also known as H56351-03
Price (USD)
Price: 158.00
Online price: 134.65
Your price:
Quantity
-

Chemical Identifiers

CAS
503-29-7
IUPAC Name
azetidine
Molecular Formula
C3H7N
InChI Key
HONIICLYMWZJFZ-UHFFFAOYSA-N
SMILES
C1CNC1
Assay (unspecified)
>97.5%

Description

Azetidine is involved in a high yielding palladium-catalyzed cross-coupling reaction with aryl bromides. Further, it is used in Ullmann type coupling reaction with iodonitroflourenes. In addition to this, it reacts with bis-(3-amino-propyl)-amine to prepare N,N'-bis-(3-amino-propyl)-propanediyldiamine using palladium as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azetidine is involved in a high yielding palladium-catalyzed cross-coupling reaction with aryl bromides. Further, it is used in Ullmann type coupling reaction with iodonitroflourenes. In addition to this, it reacts with bis-(3-amino-propyl)-amine to prepare N,N′-bis-(3-amino-propyl)-propanediyldiamine using palladium as a catalyst.

Solubility
Miscible with water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Air sensitive and heat sensitive. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text