Thermo Scientific Chemicals

4-Iodo-1H-pyrazole, 99%, Thermo Scientific Chemicals

Catalog number: H56369.06
5 g, Each
Thermo Scientific Chemicals

4-Iodo-1H-pyrazole, 99%, Thermo Scientific Chemicals

Catalog number: H56369.06
5 g, Each
Quantity
Catalog number: H56369.06
also known as H56369-06
Price (USD)
Price: 99.10
Online price: 85.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3469-69-0
IUPAC Name
4-iodo-1H-pyrazole
Molecular Formula
C3H3IN2
InChI Key
LLNQWPTUJJYTTE-UHFFFAOYSA-N
SMILES
IC1=CNN=C1
Assay (HPLC)
>98.5%

Description

4-Iodopyrazole was used in an indium-mediated synthesis of heterobiaryls. It undergoes iodination in the presence of iodine and ammonium hydroxide to yield 3,4-di-iodo- and 3,4,5-tri-iodo-pyrazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodopyrazole was used in an indium-mediated synthesis of heterobiaryls. It undergoes iodination in the presence of iodine and ammonium hydroxide to yield 3,4-di-iodo- and 3,4,5-tri-iodo-pyrazole.

Solubility
Soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text