Employed in a convenient, one-pot, synthesis of azaindoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Employed in a convenient, one-pot, synthesis of azaindoles.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
Claudio Dalvit, et al. Rapid acquisition of 1H and 19F NMR experiments for direct and competition ligand-based screening.Magn. Reson. Chem.,2011,49(4), 199-202.