D-Nipecotic acid, 96+%
D-Nipecotic acid, 96+%
D-Nipecotic acid, 96+%
Thermo Scientific Chemicals

D-Nipecotic acid, 96+%

CAS: 25137-00-2 | C6H11NO2 | 129.159 g/mol
製品番号(カタログ番号) H57434.14
または、製品番号H57434-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS25137-00-2
IUPAC Namepiperidine-3-carboxylic acid
Molecular FormulaC6H11NO2
InChI KeyXJLSEXAGTJCILF-UHFFFAOYNA-N
SMILESOC(=O)C1CCCNC1
さらに表示
Appearance (Color)White to cream
Assay from Supplier's CofA≥96.0%
FormPowder
Melting Point251-255?C
Assay from Supplier's CofAH₂O ≤ 0.5%
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only