Thermo Scientific Chemicals
4-Oxo-L-proline hydrobromide, 90+%, Thermo Scientific Chemicals
Catalog number: H57691.MD
250 mg, Each
Thermo Scientific Chemicals
4-Oxo-L-proline hydrobromide, 90+%, Thermo Scientific Chemicals
Catalog number: H57691.MD
250 mg, Each
Quantity
Catalog number: H57691.MD
also known as H57691-MD
Price (USD)
Price: 41.30
Online price: 35.65
Your price:
Quantity
-
Description
It is used as an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an active pharmaceutical intermediate.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.