2-Oxa-7-azaspiro[3.5]nonane hemioxalate, 97%
2-Oxa-7-azaspiro[3.5]nonane hemioxalate, 97%
2-Oxa-7-azaspiro[3.5]nonane hemioxalate, 97%
Thermo Scientific Chemicals

2-Oxa-7-azaspiro[3.5]nonane hemioxalate, 97%

CAS: 1429056-28-9 | C16H28N2O6 | 344.408 g/mol
数量:
250 mg
500 mg
製品番号(カタログ番号) H57834.ME
または、製品番号H57834-ME
価格(JPY)
-
数量:
500 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1429056-28-9
IUPAC Namebis(2-oxa-7-azaspiro[3.5]nonane); oxalic acid
Molecular FormulaC16H28N2O6
InChI KeyWWVUFRRXXSVWBJ-UHFFFAOYSA-N
SMILESOC(=O)C(O)=O.C1OCC11CCNCC1.C1OCC11CCNCC1
さらに表示
Assay (HPLC)96.0% min
Spiro forms of oxazines find applications as leuco dyes, by frequently displaying chromism - reversibly interchanging between their colorless and colored forms. Spiro compounds are used as photo chromic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Spiro forms of oxazines find applications as leuco dyes, by frequently displaying chromism - reversibly interchanging between their colorless and colored forms. Spiro compounds are used as photo chromic materials.

Notes
Hygroscopic. Store at room temperature.
RUO – Research Use Only

General References:

  1. Michela Bettati Dr.; Paolo Cavanni Dr.; Romano Di Fabio Dr.; Beatrice Oliosi Dr.; Ornella Perini Dr.; Gunther Scheid Dr.; Giovanna Tedesco Dr.; Laura Zonzini Dr.; Fabrizio Micheli Dr. Oxa-azaspiro Derivatives: a Novel Class of Triple Re-uptake Inhibitors. ChemMedChem. 2010, 5 (3),361-366.
  2. Erick M. Carreira.; Thomas C. Fessard. Four-Membered Ring-Containing Spirocycles: Synthetic Strategies and Opportunities.Chem. Rev. 2014, 114 (16),8257-8322.