(R)-(-)-2-Methylpyrrolidine, 95%
(R)-(-)-2-Methylpyrrolidine, 95%
(R)-(-)-2-Methylpyrrolidine, 95%
Thermo Scientific Chemicals

(R)-(-)-2-Methylpyrrolidine, 95%

CAS: 41720-98-3 | C5H11N | 85.15 g/mol
製品番号(カタログ番号) H57885.03
または、製品番号H57885-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
仕様
CAS41720-98-3
ChEBICHEBI:78858
化学物質名または材質(R)-(-)-2-Methylpyrrolidine
DOT情報Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
密度0.84
さらに表示
It is used as a building block used in a multikilogram synthesis of a chiral naphthalenoid histamine receptor antagonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a building block used in a multikilogram synthesis of a chiral naphthalenoid histamine receptor antagonist.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Keep container in a cool refrigerated, well-ventilated area. Moisture sensitive
RUO – Research Use Only

General References:

  1. Dalian Zhao.; Jeffrey T. Kuethe.; Michel Journet.; Zhihui Peng.; Guy R. Humphrey. Efficient and Practical Synthesis of (R)-2-Methylpyrrolidine.J. Org. Chem. 2006, 71 (11),4336-4338.
  2. Rumiko Sakurai.; Atsushi Yuzawa.; Kenichi Sakai.; Noriaki Hirayama. Dielectrically Controlled Enantiomeric Resolution (DCR) of (R)- and (S)-2-Methylpyrrolidine by (R,R)-Tartaric Acid.Dielectrically Controlled Enantiomeric Resolution (DCR) of (R)- and (S)-2-Methylpyrrolidine by (R,R)-Tartaric Acid. 2006, 6 (7),1606-1610.