4-(Azidomethyl)benzeneboronic acid pinacol ester, 95%
4-(Azidomethyl)benzeneboronic acid pinacol ester, 95%
4-(Azidomethyl)benzeneboronic acid pinacol ester, 95%
Thermo Scientific Chemicals

4-(Azidomethyl)benzeneboronic acid pinacol ester, 95%

CAS: 1239481-05-0 | C13H18BN3O2 | 259.116 g/mol
製品番号(カタログ番号) H58401.03
または、製品番号H58401-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1239481-05-0
IUPAC Name2-[4-(azidomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular FormulaC13H18BN3O2
InChI KeyCARNXRHOIGMOQD-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC=C(CN=[N+]=[N-])C=C1
さらに表示
Assay from Supplier's CofA≥94.0%
Appearance (Color)Colorless to white
FormSolid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Incompatible with oxidizing agents. Store in a dry, cool conditions in a well sealed containers. Store at 4°C.
RUO – Research Use Only