1-Mercaptoadamantane, 98%
1-Mercaptoadamantane, 98%
1-Mercaptoadamantane, 98%
Thermo Scientific Chemicals

1-Mercaptoadamantane, 98%

CAS: 34301-54-7 | C10H16S | 168.298 g/mol
製品番号(カタログ番号) H58958.MD
または、製品番号H58958-MD
価格(JPY)
-
数量:
250 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS34301-54-7
IUPAC Nameadamantane-1-thiol
Molecular FormulaC10H16S
InChI KeyADJJLNODXLXTIH-UHFFFAOYSA-N
SMILESSC12CC3CC(CC(C3)C1)C2
さらに表示
Appearance (Color)White to pale cream
FormCrystalline powder or powder
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only