Thermo Scientific Chemicals

L-Phenylalanine tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H59122.06
5 g, Each
Thermo Scientific Chemicals

L-Phenylalanine tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H59122.06
5 g, Each
Quantity
Catalog number: H59122.06
also known as H59122-06
Price (USD)
Price: 184.00
Online price: 158.65
Your price:
Quantity
-

Chemical Identifiers

CAS
15100-75-1
IUPAC Name
hydrogen tert-butyl 2-amino-3-phenylpropanoate chloride
Molecular Formula
C13H20ClNO2
InChI Key
FDMCEXDXULPJPG-UHFFFAOYNA-N
SMILES
[H+].[Cl-].CC(C)(C)OC(=O)C(N)CC1=CC=CC=C1
Assay (HPLC)
>97.5%

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and food industry. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and food industry. 

Solubility
Soluble in methanol.

Notes
Hygroscopic. Keep away from heat and humidity. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text