N-Boc-O-tert-butyl-L-serine dicyclohexylammonium salt, 98%
N-Boc-O-tert-butyl-L-serine dicyclohexylammonium salt, 98%
N-Boc-O-tert-butyl-L-serine dicyclohexylammonium salt, 98%
N-Boc-O-tert-butyl-L-serine dicyclohexylammonium salt, 98%
Thermo Scientific Chemicals

N-Boc-O-tert-butyl-L-serine dicyclohexylammonium salt, 98%

CAS: 18942-50-2 | C24H46N2O5 | 442.641 g/mol
製品番号(カタログ番号) H59163.06
または、製品番号H59163-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS18942-50-2
IUPAC Name3-(tert-butoxy)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid; N-cyclohexylcyclohexanamine
Molecular FormulaC24H46N2O5
InChI KeyAIEUUHIXSUNTGV-UHFFFAOYNA-N
SMILESC1CCC(CC1)NC1CCCCC1.CC(C)(C)OCC(NC(=O)OC(C)(C)C)C(O)=O
さらに表示
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only