Ethyl 6-bromo-4-chloroquinoline-3-carboxylate, 97%, Thermo Scientific Chemicals
Ethyl 6-bromo-4-chloroquinoline-3-carboxylate, 97%, Thermo Scientific Chemicals
Ethyl 6-bromo-4-chloroquinoline-3-carboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 6-bromo-4-chloroquinoline-3-carboxylate, 97%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
H59301.03
または、製品番号H59301-03
1 g
製品番号(カタログ番号) H59301.03
または、製品番号H59301-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
仕様
CAS206257-39-8
化学物質名または材質Ethyl 6-bromo-4-chloroquinoline-3-carboxylate
式量314.57
健康被害1H315-H319-H335
健康被害2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
さらに表示
Ethyl 6-bromo-4-chloroquinoline-3-carboxylate is used as pharmaceutical intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 6-bromo-4-chloroquinoline-3-carboxylate is used as pharmaceutical intermediates

Solubility
Immiscible in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Qingsong Liu.; Jinhua Wang.; Seong A. Kang.; Carson C. Thoreen.; Wooyoung Hur.; Tausif Ahmed.; David M. Sabatini.; Nathanael S. Gray. Discovery of 9-(6-Aminopyridin-3-yl)-1-(3-(trifluoromethyl)phenyl)benzo[h][1,6]naphthyridin-2(1H)-one (Torin2) as a Potent, Selective, and Orally Available Mammalian Target of Rapamycin (mTOR) Inhibitor for Treatment of Cancer. J. Med. Chem. 2011, 54, (5), 1473-1480.