2-Amino-3,6-difluorobenzonitrile, 97%
2-Amino-3,6-difluorobenzonitrile, 97%
2-Amino-3,6-difluorobenzonitrile, 97%
2-Amino-3,6-difluorobenzonitrile, 97%
Thermo Scientific Chemicals

2-Amino-3,6-difluorobenzonitrile, 97%

CAS: 190011-81-5 | C7H4F2N2 | 154.12 g/mol
製品番号(カタログ番号) H59362.03
または、製品番号H59362-03
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
仕様
CAS190011-81-5
化学物質名または材質2-Amino-3,6-difluorobenzonitrile
DOT情報Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
式量154.12
健康被害1H302+H312+H332-H315-H319-H335
さらに表示
2-Amino-3,6-difluorobenzonitrile is used in cosmetic and pharmaceutical intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-3,6-difluorobenzonitrile is used in cosmetic and pharmaceutical intermediates

Solubility
Miscible with water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only