1-Ethyl-3-methylimidazolium iodide, 97%
1-Ethyl-3-methylimidazolium iodide, 97%
1-Ethyl-3-methylimidazolium iodide, 97%
Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium iodide, 97%

CAS: 35935-34-3 | C6H11IN2 | 238.07 g/mol
製品番号(カタログ番号) H59404.06
または、製品番号H59404-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS35935-34-3
Appearance (Color)White to pale yellow
Proton NMR>96.0%
FormPowder
It is used as OLED materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as OLED materials.

Solubility
Soluble in water, , dichloromethane, ethanol, acetonitrile.

Notes
Moisture Sensitive. Keep away from heat and humidity. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Annegret Stark.; Bonnie L. MacLean.; Robert D. Singer. 1-Ethyl-3-methylimidazolium halogenoaluminate ionic liquids as solvents for Friedel-Crafts acylation reactions of ferrocene. J. Chem. Soc., Dalton Trans. 1999, (1),63-66.
  2. Laine Green.; Ivan Hemeon.; Robert D Singer. 1-Ethyl-3-methylimidazolium halogenoaluminate ionic liquids as reaction media for the acylative cleavage of ethers.Tetrahedron Lett. 2000, 41 (9), 1343-1346.